Learn More
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. Azlon™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
Brand: Quickfit SB34
109.25 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.30600kg
Description
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. AzlonTM Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
- Conforms to the standard tapers as outlined in ISO 383
- Borosilicate glass for chemical durability and high temperature performance
- Hollow-blown construction to reduce weight
- Hexagonal head prevents the stopper from rolling and provides a good grip
- Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Specifications
Stopper | |
Borosilicate Glass | |
Standard; 34/35 |
Chemical durability and high temperature performance | |
Quickfit™ jointed ware and other Glassware having standard ground Glass sockets |