Learn More
Quickfit™ Straight Tubing Adapter
Click to view available options
Standard Taper Joint No.:
24/29
29/32
Unit Size:
Pack of 5
Description
- With straight connection for the attachment of rubber hose etc
- Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
Specifications
Specifications
| For Use With (Equipment) | Flexible tubing, Rubber hose |
| Includes | Detachable Plastic screw thread connector |
| Material | Borosilicate Glass |
| Product Type | Straight tubing adapter |
| Standard Taper Joint No. | 24/29 |