Learn More
Quickfit™ Straight Tubing Adapter
Product Code. 15547930
Click to view available options
Standard Taper Joint No.:
24/29
29/32
Unit Size:
Pack of 5
Product Code. 15547930
Brand: Quickfit MF13/4/SC
Only null left
Add to basket
Add to basket
Add detachable tubing to a glass vescle with a glass tubing adapter. QuickFit™ straight tubing adapters come in different sizes to fit containers or varying sizes.
  • With straight connection for the attachment of rubber hose etc
  • Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Flexible tubing, Rubber hose
Includes Detachable Plastic screw thread connector
Material Borosilicate Glass
Product Type Straight tubing adapter
Standard Taper Joint No. 29/32