Learn More
Quickfit™ Vacuum Receiver Adapter
Product Code. 11312105
Click to view available options
Height (Metric):
110mm
140mm
150mm
165mm
Cone Size:
14/23
19/26
24/29
29/32
Standard Taper Joint No.:
19/26
24/29
Unit Size:
Each
Product Code. 11312105
Brand: Quickfit RA3/44/SC
Only null left
Add to basket
Add to basket
Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.
Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.
  • Delivery stem within cone
  • With sidearm for connection to a vacuum supply
  • Easy connection to flexible tubing via detachable plastic screwthread connector
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 29/32
For Use With (Equipment) Flexible tubing
Height (English) 6.49in.
Height (Metric) 165mm
Includes Detachable Plastic screw thread connector, delivery stem, side-arm
Material Borosilicate Glass
Product Type Vacuum receiver angled adapter
Standard Taper Joint No. 24/29