missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Quinidine Monoclonal Antibody (QD11-261.3.6)

Description
MA1-10747 detects Quinidine. MA1-10747 has been successfully used in ELISA applications. The MA1-10747 immunogen is Quinidine, conjugated to KLH.
Quinidine is an alkaloid extracted from the bark of the Cinchona tree with class 1A antiarrhythmic and antimalarial effects. Quinidine stabilizes the neuronal membrane by binding to and inhibiting voltage-gated sodium channels, thereby inhibiting the sodium influx required for the initiation and conduction of impulses resulting in an increase of the threshold for excitation and decreased depolarization during phase 0 of the action potential. In addition, the effective refractory period (ERP), action potential duration (APD), and ERP/APD ratios are increased, resulting in decreased conduction velocity of nerve impulses. Quinidine exerts its antimalarial activity by acting primarily as an intra-erythrocytic schizonticide through association with the heme polymer (hemazoin) in the acidic food vacuole of the parasite thereby preventing further polymerization by heme polymerase enzyme. This results in accumulation of toxic heme and death of the parasite. [National Center for Biotechnology Information. PubChem Compound Database; CID=441074].
Specifications
Specifications
| Antigen | Quinidine |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | QD11-261.3.6 |
| Concentration | 1.08 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | C20H24N2O2; Chinidin; Cin-Quin; Pitayine; Quinidex |
| Host Species | Mouse |
| Immunogen | Quinidine, conjugated to KLH. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?