missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4509880
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
486-74-8
C10H7NO2
173.171
VQMSRUREDGBWKT-UHFFFAOYSA-N
4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure
10243
CHEBI:18311
quinoline-4-carboxylic acid
C1=CC=C2C(=C1)C(=CC=N2)C(=O)O
This item is not returnable. View return policy
486-74-8
C10H7NO2
173.171
VQMSRUREDGBWKT-UHFFFAOYSA-N
4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure
10243
CHEBI:18311
quinoline-4-carboxylic acid
C1=CC=C2C(=C1)C(=CC=N2)C(=O)O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material Quinoline-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'