Learn More
Quinoxaline-6-boronic acid pinacol ester, 97%, Thermo Scientific™
Catalog Number p-4380063
Click to view available options
Quantity:
1 g
5 g
1167418-13-4
C14H17BN2O2
256.11
MFCD11054040
ZYWICCYXTGRUNM-UHFFFAOYSA-N
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid, pinacol ester, 6-quinoxaline boronic acid pinacol ester, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
44118298
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline
CC1(C)OB(OC1(C)C)C1=CC2=NC=CN=C2C=C1
1167418-13-4
C14H17BN2O2
256.11
MFCD11054040
ZYWICCYXTGRUNM-UHFFFAOYSA-N
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid, pinacol ester, 6-quinoxaline boronic acid pinacol ester, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
44118298
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline
CC1(C)OB(OC1(C)C)C1=CC2=NC=CN=C2C=C1
Boiling Point 130°C to 145°C (0.6 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 256.11
Percent Purity 97%
Chemical Name or Material Quinoxaline-6-boronic acid pinacol ester
compliance-icons
Product Identifier
  • Quinoxaline-6-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.