missing translation for 'onlineSavingsMsg'
Learn More

Quisinostat, MedChemExpress

Product Code. 16479097
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479097 5 mg 5mg
16466766 100 mg 100mg
16446518 10 mg 10mg
16412617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16479097 Supplier Medchem Express Supplier No. HY15433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quisinostat (JNJ-26481585) is a potent, second-generation and orally active pan-HDAC inhibitor (HDACi), with IC50 values ranging from 0.11 nM to 0.64 nM for HDAC1, HDAC2, HDAC4, HDAC10 and HDAC11. Quisinostat has a broad spectrum antitumoral activity. Quisinostat can induce autophagy in neuroblastoma cells.

Quisinostat (JNJ-26481585) is a potent, second-generation and orally active pan-HDAC inhibitor (HDACi), with IC50 values ranging from 0.11 nM to 0.64 nM for HDAC1, HDAC2, HDAC4, HDAC10 and HDAC11. Quisinostat has a broad spectrum antitumoral activity. Quisinostat can induce autophagy in neuroblastoma cells.

Specifications

Chemical Name or Material Quisinostat
CAS 875320-29-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H26N6O2
Quantity 5 mg
Synonym JNJ-26481585
Solubility Information DMSO : 50 mg/mL (126.75 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(N2CCC(CNCC3=CN(C)C4=C3C=CC=C4)CC2)N=C1)NO
Molecular Weight (g/mol) 394.47
Formula Weight 394.47
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.