Learn More
QX77, MedChemExpress
Product Code. 16490927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16490927
Brand: Medchem Express HY112483
MedChemExpress QX77 is a chaperone-mediated autophagy (CMA) activator and upregulates LAMP2A expression in vitro. QX77 induces Rab11 upregulation, rescues Rab11 down-regulation and trafficking deficiency in cystinotic cells. QX77 can impede self-renewal and promote differentiation of ES cells.
QX77 is a chaperone-mediated autophagy (CMA) activator and upregulates LAMP2A expression in vitro. QX77 induces Rab11 upregulation, rescues Rab11 down-regulation and trafficking deficiency in cystinotic cells. QX77 can impede self-renewal and promote differentiation of ES cells.
Specifications
Chemical Name or Material QX77
CAS 1798331-92-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H13ClN2O2
Quantity 50 mg
Solubility Information DMSO : 15 mg/mL (49.88 mM; ultrasonic and warming and heat to 60°C)
SMILES CC(NC1=CC=C(C2=NC3=CC=C(Cl)C=C3OC2)C=C1)=O
Molecular Weight (g/mol) 300.74
Formula Weight 300.74
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only