missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine, 95%

Brand:  Alfa Aesar™ H64965.06

Additional Details : CAS : 100858-33-1 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15439286

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

100858-33-1
221.256
MBLJFGOKYTZKMH-LLVKDONJSA-N
11183628
C1CN(CC1O)C(=O)OCC2=CC=CC=C2
C12H15NO3
MFCD07368258
r---1-cbz-3-pyrrolidinol, benzyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-1-cbz-3-pyrrolidinol, r-benzyl 3-hydroxypyrrolidine-1-carboxylate, r-n-cbz-3-hydroxypyrrolidine, r-1-carbobenzoxy-3-pyrrolidinol, r-1-cbzl-3-pyrrolidinol, r-n-z-3-pyrrolidinol, r---1-cbz-3-hydroxy-pyrrolidine, 3r-n-cbz-3-pyrrolidinol
benzyl (3R)-3-hydroxypyrrolidine-1-carboxylate

Specifications

(R)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine
5g
C12H15NO3
UN2811
r---1-cbz-3-pyrrolidinol, benzyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-1-cbz-3-pyrrolidinol, r-benzyl 3-hydroxypyrrolidine-1-carboxylate, r-n-cbz-3-hydroxypyrrolidine, r-1-carbobenzoxy-3-pyrrolidinol, r-1-cbzl-3-pyrrolidinol, r-n-z-3-pyrrolidinol, r---1-cbz-3-hydroxy-pyrrolidine, 3r-n-cbz-3-pyrrolidinol
C1CN(CC1O)C(=O)OCC2=CC=CC=C2
221.256
221.26
95%
74°C to 79°C
100858-33-1
MFCD07368258
5016071
MBLJFGOKYTZKMH-LLVKDONJSA-N
benzyl (3R)-3-hydroxypyrrolidine-1-carboxylate
11183628
Solid
Videos
Documents