Learn More
BUCHI R-100 System
Entry level Rotavapor to meet the essential needs in evaporation and BUCHI’s high quality standards.
Brand:  BUCHI 11100V1021
Additional Details : Weight : 53.60000kg
 View more versions of this product
Product Code. 15364906
  • € 9665.00 / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • R-100 rotary evaporator
  • Entry level Rotavapor
  • Meets the essential needs in evaporation and BUCHI’s high quality standards.
Specifications
Specifications
No Protective coating
5 L
Ambient to 95°C
Vertical
220/240 V
280 rpm
F -105, -10°C to +25°C
SJ 29/32