missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress R-1479
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Molecular Weight (g/mol) | 284.23 g/mol |
| Product Type | Reference compound |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 98.66% |
| Quantity | 100 mg |
| Solubility Information | DMSO : 100 mg/mL (351.83 mM∣Need ultrasonic) |
| For Use With (Application) | COVID-19-anti-virus |
| CAS | 478182-28-4 |
| Synonym | 4'-Azidocytidine |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction