Learn More
(R)-2-Amino-N-benzyl-3-methoxypropionamide, TRC
Product Code. 30138660
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138660 1 g 1g
30134108 250 mg 250mg
30004362 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138660 Supplier Toronto Research Chemicals Supplier No. TRCA5980001G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 196601-69-1
Molecular Formula C11 H16 N2 O2
Molecular Weight (g/mol) 208.26
Synonym Propanamide, 2-amino-3-methoxy-N-(phenylmethyl)-, (R)-, (2R)-2-Amino-3-methoxy-N-(phenylmethyl)propanamide, (2R)-2-Amino-3-methoxy-N-(phenylmethyl)propanamide, (2R)-2-Amino-N-benzyl-3-methoxypropanamide, (R)-2-Amino-N-benzyl-3-methoxypropionamide
IUPAC Name (2R)-2-amino-N-benzyl-3-methoxypropanamide
SMILES COC[C@@H](N)C(=O)NCc1ccccc1
Specifications
Analyte or Component Names (R)-2-Amino-N-benzyl-3-methoxypropionamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H16N2O2/c1-15-8-10(12)11(14)13-7-9-5-3-2-4-6-9/h2-6,10H,7-8,12H2,1H3,(H,13,14)/t10-/m1/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 1 g
Formula Weight 208.1212
Percent Purity >95%
Chemical Name or Material (R)-2-Amino-N-benzyl-3-methoxypropionamide
Grade HPLC