missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Methylmorpholine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7077637
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
953780-78-4
C5H11NO·ClH
137.61
MSOCQCWIEBVSLF-NUBCRITNSA-N
r-3-methylmorpholine hydrochloride, 3r-3-methylmorpholine hydrochloride, c5h11no.clh, ksc494m2h, r-3-methylmorpholin hydrochloride, r-3-methylmorpholinehydrochloride, r-3-methyl-morpholine hydrochloride, 3r-3-methyl-morpholine hydrochloride, 3r-3-methylmorpholine-hydrogen chloride 1/1
57356922
(3R)-3-methylmorpholine;hydrochloride
CC1COCCN1.Cl
This item is not returnable. View return policy
953780-78-4
C5H11NO·ClH
137.61
MSOCQCWIEBVSLF-NUBCRITNSA-N
r-3-methylmorpholine hydrochloride, 3r-3-methylmorpholine hydrochloride, c5h11no.clh, ksc494m2h, r-3-methylmorpholin hydrochloride, r-3-methylmorpholinehydrochloride, r-3-methyl-morpholine hydrochloride, 3r-3-methyl-morpholine hydrochloride, 3r-3-methylmorpholine-hydrogen chloride 1/1
57356922
(3R)-3-methylmorpholine;hydrochloride
CC1COCCN1.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 126.0°C to 128.5°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Quantity 1 g
Specific Rotation 1.86
Formula Weight 137.61
Percent Purity 97%
Chemical Name or Material (R)-3-Methylmorpholine hydrochloride
compliance-icons
Product Identifier
  • (R)-3-Methylmorpholine hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'