Learn More
(R)-5,5',6,6',7,7',8,8'-Octahydro[1,1'-binaphthalene]-2,2'-diol, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 65355-14-8 |
| Molecular Formula | C20H22O2 |
| MDL Number | MFCD02093485 |
| Quantity | 100 g |
| SMILES | OC1=CC=C2CCCCC2=[C@]1[C@]3=C4CCCCC4=CC=C3O |
| Molecular Weight (g/mol) | 294.39 |
| Percent Purity | 97% |
| Chemical Name or Material | (R)-5,5',6,6',7,7',8,8'-Octahydro[1,1'-binaphthalene]-2,2'-diol |