missing translation for 'onlineSavingsMsg'
Learn More

R-7050, MedChemExpress

Product Code. 16483046
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483046

Brand: Medchem Express HY110203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R-7050 (TNF-α Antagonist III) is a tumor necrosis factor receptor (TNFR) antagonist with greater selectivity toward TNFα.

R-7050 (TNF-α Antagonist III) is a tumor necrosis factor receptor (TNFR) antagonist with greater selectivity toward TNFα.

Specifications

Chemical Name or Material R-7050
Physical Form Solid
CAS 303997-35-5
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H8ClF3N4S
For Use With (Application) Cancer-Kinase/protease
Synonym TNF-α Antagonist III
Solubility Information DMSO : 25 mg/mL (65.66 mM; Need ultrasonic)
SMILES FC(F)(C1=NN=C2N1C3=CC(Cl)=CC=C3N=C2SC4=CC=CC=C4)F
Molecular Weight (g/mol) 380.77
Formula Weight 380.77
Percent Purity 99.26%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.