Learn More
(R)-Afatinib, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-Afatinib |
| CAS | 945553-91-3 |
| Molecular Formula | C24H25ClFN5O3 |
| Quantity | 10 mg |
| Synonym | (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[[(3R)-tetrahydro-3-furanyl]oxy]-6-quinazolinyl]-4-(dimethylamino)-2-butenamide, ent-Afatinib |
| InChI Formula | InChI=1S/C24H25ClFN5O3/c1-31(2)8-3-4-23(32)30-21-11-17-20(12-22(21)34-16-7-9-33-13-16)27-14-28-24(17)29-15-5-6-19(26)18(25)10-15/h3-6,10-12,14,16H,7-9,13H2,1-2H3,(H,30,32)(H,27,28,29)/b4-3+/t16-/m1/s1 |
| SMILES | O=C(NC1=CC2=C(NC3=CC=C(F)C(Cl)=C3)N=CN=C2C=C1O[C@H]4COCC4)/C=C/CN(C)C |
| IUPAC Name | (E)-N-[4-(3-chloro-4-fluoroanilino)-7-[(3R)-oxolan-3-yl]oxyquinazolin-6-yl]-4-(dimethylamino)but-2-enamide |
| Molecular Weight (g/mol) | 485.94 |
| Formula Weight | 485.163 |