Learn More
(R)-(+)-alpha,alpha-Diphenylprolinol, 99%, Thermo Scientific™
Product Code. 11347296
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 11347296
Brand: Thermo Scientific Alfa Aesar L09218.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22348-32-9
Molecular Formula C17H19NO
Molecular Weight (g/mol) 253.345
MDL Number MFCD00077754
InChI Key OGCGXUGBDJGFFY-MRXNPFEDSA-N
Synonym r-diphenyl pyrrolidin-2-yl methanol, r-+-alpha,alpha-diphenyl-2-pyrrolidinemethanol, r-+-2-diphenylhydroxymethyl pyrrolidine, diphenyl 2r-pyrrolidin-2-ylmethanol, r-+-2-diphenylhydroxymethyl-pyrrolidin, diphenyl-2r-pyrrolidin-2-yl methanol, r-alpha,alpha-diphenyl-2-pyrrolidinemethanol, alpha,alpha-diphenyl-d-prolinol, r-+-alpha,alpha-diphenylprolinol, diphenyl 2r-pyrrolidin-2-yl methanol
PubChem CID 7045371
IUPAC Name diphenyl-[(2R)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O
Specifications
Melting Point 76°C to 78°C
Quantity 250 mg
Optical Rotation +57.5° (c=3 in Methanol)
Beilstein 4353363
Formula Weight 253.35
Percent Purity 99%
Chemical Name or Material (R)-(+)-alpha,alpha-Diphenylprolinol