missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cerestat

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Cerestat is a potent and noncompetitive NMDA receptor antagonist (IC50 = 36 nM). Exhibits >70-fold selectivity for NMDA receptor over σ receptor. Neuroprotective in a rat model of focal ischemia.
Specifications
Specifications
| Chemical Name or Material | Cerestat |
| CAS | 137160-11-3 |
| Synonym | N-(3-Ethylphenyl)-N-methyl-N'-1-naphthalenylguanidine hydrochloride |
| Quantity | 50 mg |
| Solubility | Soluble to 50mM in water and to 100mM in DMSO |
| Molecular Formula | C20H21N3·HCl |
| Formula Weight | 339.86 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction