missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Echinomycin

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Echinomycin is a highly potent and selective HIF-1α inhibitor (IC50 = 29.4 pM). Selectively inhibits HIF-1 binding to the VEGF promoter without affecting the binding of AP-1 or NF-κB. Inhibits colony formation of cancer stem cells (CSC) with a 100-fold selectivity over normal hematopoietic progenitor cells. Eradicates mouse lymphomas and human AML xenografts by eliminating CSCs.
Specifications
Specifications
| Chemical Name or Material | Echinomycin |
| CAS | 512-64-1 |
| Synonym | N-(2-Quinoxalinylcarbonyl)-O-[N-(2-quinoxalinylcarbonyl)-D-seryl-L-alanyl-3-mercapto-N,S-dimethylcysteinyl-N-methyl-L-valyl]-D-seryl-L-alanyl-N-methylcysteinyl-N-methyl-L-valine-(81)-lactone-cyclic (37)-thioether |
| Quantity | 1 mg |
| Solubility | Soluble to 5mM in DMSO |
| Molecular Formula | C51H64N12O12S2 |
| Formula Weight | 1101.26 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction