missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flow Cytometry Fixation/Permeabilization Buffer I (1X), R&D Systems™
Description
- Designed for intracellular flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies
Applications:
Flow Cytometry
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C for up to 6 months from date of receipt. |
| Formulation | Sodium azide 0.05% and formaldehyde 4% |
| Quantity | 125mL |
| Stability | Upto 6 Months |
| Product Type | Fixation/Permeabilization Buffer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?