missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flow Cytometry Fixation and Permeabilization Buffer Kit I, R&D Systems™
Description
- Designed for intracellular flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies
- Includes 100mL of Flow Cytometry Fixation Buffer (1X) and 250mL of Flow Cytometry Permeabilization/Wash Buffer I (1X)
Applications:
Flow Cytometry
Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Formulation | Sodium azide 0.05% and formaldehyde 4% |
| Includes | 100mL flow cytometry fixation buffer 1X, 250mL flow cytometry permeabilization/wash buffer I 1X |
| Quantity | 1 Kit |
| Stability | Upto 6 Months |
| Product Type | Fixation/Permeabilization Buffer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?