missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GlyT1/SLC6A9 (NP_964012) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Human GlyT1/SLC6A9 (NP_964012) VersaClone cDNA clone contains the complete ORF for GlyT1/SLC6A9, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GlyT1/SLC6A9 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
| Product Type | cDNA |
| Gene | hSLC6A9 |
| Accession Number | NP_964012 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 2134bp |
| Gene ID (Entrez) | 6536, 14664, 116509 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human, Mouse, Rat |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction