Learn More
Tocris Bioscience™ IPTG
Used in cloning procedures with X-GAL
Brand: Tocris Bioscience™ 5509/10G
Description
IPTG is an used in combination with X-GAL (Cat. No. 5510) in cloning procedures requiring expression of β-galactosidase activity. Non-metabolizable analog of galactose.This product has <0.1% dioxane content.
Specifications
| 1-Methylethyl 1-thio-β-D-galactopyranoside | |
| 1-Methylethyl 1-thio-β-D-galactopyranoside | |
| Soluble to 100mM in water | |
| Bioactive Small Molecule |
| 367-93-1 | |
| 10 g | |
| Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research Use Only. Not For Human or Veterinary Use