missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IFN-gamma ELISpot Kit
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
Description
Product Features:
- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- Hig
Specifications
Specifications
| Includes | Six 96-well PVDF-backed plates coated w/mAb for mouse IFNG, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control |
| Quantity | 6 x 96-well Plates |
| Sample Type | Whole Cells |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Diagnostics |
| Product Type | Mouse IFN-gamma ELISpot Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction