missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Trypsin 3/PRSS3 Protein, CF

Product Code. 18168280
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18168280

Brand: R&D Systems™ 3565SE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >17,000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_035775
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HCl and NaCl.
Gene ID (Entrez) 5646
Molecular Weight (g/mol) M.W. (Observed): 29 kDa and 36 kDa, reducing conditions; M.W. (theoretical): 26 kDa
Name Trypsin 3/PRSS3
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse Trypsin 3/PRSS3 protein Phe16-Asn246, with a C-terminal 10-His tag Accession # NP_035775
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Structural Form Pro form
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Prss3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Trypsin 3/PRSS3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.