missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PR 619

Description
PR 619 is a broad spectrum, reversible DUB inhibitor. Exhibits limited activity against other proteases. Induces accumulation of polyubiquitinated proteins, but has no direct inhibitory effect on the proteasome. Cytotoxic in HEK293T and colorectal cancer cells. Stabilizes microtubule network in oligodendroglial cells.
Specifications
Specifications
| Chemical Name or Material | PR 619 |
| CAS | 2645-32-1 |
| Synonym | 2,6-Diamino-3,5-dithiocyanopyridine |
| Quantity | 50 mg |
| Solubility | Soluble to 10mM in DMSO |
| Molecular Formula | C7H5N5S2 |
| Formula Weight | 223.28 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?