Learn More
R&D Systems™ Recombinant Human GDF-9
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 8266-G9-010
Description
The Recombinant Human GDF-9 Protein is derived from HEK293. The Recombinant Human GDF-9 Protein has been validated for the following applications: Bioactivity.Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately
Specifications
| O60383 | |
| 16 kDa (monomer) | |
| 10μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| Yes |
| Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. | |
| GDF-9 | |
| Human embryonic kidney cell,HEK293-derived human GDF-9 protein Gly320-Arg454 (Gly391Arg) | |
| Unconjugated |