Learn More
R&D Systems™ Sialyltransferase Activity Kit
Product Code. 18712114
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18712114 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18712114 Supplier R&D Systems™ Supplier No. EA002
Includes
CHO cell-expressed Recombinant Human CD73, CMP, Phosphatase Buffer 2, 100mM MnCl2, Phosphate Standard, Malachite Green Reagents A and B
A simple, nonradioactive high throughput-compatible format for assaying the enzyme activity of all sialyltransferases regardless of different acceptor substrates
Uses the 5 ft.-nucleotidase CD73 as a coupling phosphatase to remove inorganic phosphate quantitatively from the leaving nucleotide cytidine 5 ft.-monophosphate (CMP) that is generated during sialyltransferase reactions
No t
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 Kit