missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Epichlorohydrin, 99%

Product Code. p-3778519 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193331 1 g Glass bottle 1g
10076303 5 g Glass bottle 5g
10341534 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193331 Supplier Thermo Scientific Acros Supplier No. 349010010
Add to basket
This item is not returnable. View return policy

CAS: 51594-55-9 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51594-55-9
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00077759
InChI Key BRLQWZUYTZBJKN-VKHMYHEASA-N
Synonym r---epichlorohydrin, r-epichlorohydrin, 2r-2-chloromethyl oxirane, --epichlorohydrin, r-chloromethyl oxirane, r-2-chloromethyl oxirane, epichlorohydrin,-, unii-vec01h609i, r-1-chloro-2,3-epoxypropane
PubChem CID 2734062
ChEBI CHEBI:18662
IUPAC Name (2R)-2-(chloromethyl)oxirane
SMILES ClC[C@H]1CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1830g/mL
Boiling Point 114°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4365 to 1.4385
Quantity 1 g
Specific Gravity 1.183
Specific Rotation Condition +34° (20°C 589nm,c=1,MeOH)
Specific Rotation + 34
Solubility Information Solubility in water: insoluble in water
Formula Weight 92.52
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (R)-(-)-Epichlorohydrin
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-(-)-Epichlorohydrin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.