Learn More
R&D Systems™ Human Integrin beta 4/CD104 (NP_000204) VersaClone cDNA
Product Code. 18705918
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 18705918
Brand: R&D Systems™ RDC1079
CD104; ITGB4
The Human Integrin beta 4/CD104 (NP_000204) VersaClone cDNA clone contains the complete ORF for Integrin beta 4/CD104, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Integrin beta 4/CD104 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans