Learn More
(R)-Methotrexate, TRC
Specifications
Specifications
| Chemical Name or Material | (R)-Methotrexate |
| CAS | 51865-79-3 |
| Molecular Formula | C20H22N8O5 |
| Quantity | 1 mg |
| Synonym | (R)-2-(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}benzamido)pentanedioic acid |
| InChI Formula | InChI=1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/t13-/m1/s1 |
| SMILES | CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@H](CCC(=O)O)C(=O)O |
| IUPAC Name | (2R)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid |
| Molecular Weight (g/mol) | 454.54 |
| Formula Weight | 454.1713 |