missing translation for 'onlineSavingsMsg'
Learn More

(R)-α-Methyltyrosine, 98%, 98% ee, Thermo Scientific™

Catalog Number p-4259043
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
672-86-6
C10H13NO3
195.22
NHTGHBARYWONDQ-SNVBAGLBSA-N
r-alpha-methyltyrosine, alpha-methyl-d-tyr, r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, 2r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, alpha-methyl-d-tyrosine, metirosinum inn-latin, metirosina inn-spanish, l-tyrosine, alpha-methyl-,-, d +-metyrosine, metyrosine usan
12650
(2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
CC(CC1=CC=C(C=C1)O)(C(=O)O)N
This item is not returnable. View return policy
672-86-6
C10H13NO3
195.22
NHTGHBARYWONDQ-SNVBAGLBSA-N
r-alpha-methyltyrosine, alpha-methyl-d-tyr, r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, 2r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, alpha-methyl-d-tyrosine, metirosinum inn-latin, metirosina inn-spanish, l-tyrosine, alpha-methyl-,-, d +-metyrosine, metyrosine usan
12650
(2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
CC(CC1=CC=C(C=C1)O)(C(=O)O)N
CAS Min % 97.5
CAS Max % 100.0
Melting Point >211°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Specific Rotation -18
Formula Weight 195.22
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-α-Methyltyrosine, 98% ee
Show More Show Less
Product Identifier
  • (R)-alpha-Methyltyrosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'