Learn More
(R)-(-)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate, 95%, Thermo Scientific™
Product Code. 11441130
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441130 1 g 1g
11451130 5 g 5g
11461130 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11441130 Supplier Thermo Scientific Alfa Aesar Supplier No. H25778.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C18H25FeNO6
MDL Number MFCD09953453
Specifications
Melting Point 129°C to 131°C
Quantity 1 g
Optical Rotation +14° (c=1 in Methanol)
Formula Weight 407.26
Percent Purity 95%
Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate