Learn More
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals
Product Code. 11355505
Click to view available options
Quantity:
100 mg
500 mg
2 g
Unit Size:
100mg
2g
500mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11355505
Brand: Thermo Scientific Alfa Aesar L19685.04
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55700-44-2
Molecular Formula C26H28FeNP
Molecular Weight (g/mol) 441.34
MDL Number MFCD00075286
InChI Key RCAFPSZHKFOLEE-KVEBOLLKNA-N
IUPAC Name (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
SMILES [Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 141°C to 143°C
Quantity 2 g
Optical Rotation −355° (c=0.6 in Ethanol)
Formula Weight 441.34
Percent Purity 97%
Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.