Learn More
(R/S)-N-Deacetyl Colchiceine N-Trifluroracetate, TRC
Specifications
Specifications
| Chemical Name or Material | (R/S)-N-Deacetyl Colchiceine N-Trifluroracetate |
| CAS | 102491-70-3 |
| Molecular Formula | C21H20F3NO6 |
| Quantity | 25 mg |
| Synonym | 2,2,2-Trifluoro-N-[5,6,7,9-tetrahydro-10-hydroxy-1,2,3-trimethoxy-9-oxobenzo[a]heptalen-7-yl]-acetamide |
| InChI Formula | InChI=1S/C21H20F3NO6/c1-29-16-8-10-4-6-13(25-20(28)21(22,23)24)12-9-15(27)14(26)7-5-11(12)17(10)19(31-3)18(16)30-2/h5,7-9,13H,4,6H2,1-3H3,(H,25,28)(H,26,27) |
| SMILES | COc1cc2CCC(NC(=O)C(F)(F)F)C3=CC(=O)C(=CC=C3c2c(OC)c1OC)O |
| IUPAC Name | 2,2,2-trifluoro-N-(10-hydroxy-1,2,3-trimethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl)acetamide |
| Molecular Weight (g/mol) | 439.38 |
| Formula Weight | 439.1243 |
Product Title
Ser du en mulighed for forbedring?