Learn More
Novus Biologicals™ R-Spondin 1 Antibody (Oncomed patent anti-RSPO1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28641-50ug
 View more versions of this product
Product Code. 30046777
  • € 213.00 / 50µg
Description
R-Spondin 1 Monoclonal antibody specifically detects R-Spondin 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
R-Spondin 1
Monoclonal
Unconjugated
Q2MKA7
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Oncomed patent anti-RSPO1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
RSPO1
50 μg
Signal Transduction
284654