Learn More
Novus Biologicals™ R-Spondin 1 Antibody (Oncomed patent anti-RSPO1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28641-100ug
This item is not returnable.
View return policy
Description
R-Spondin 1 Monoclonal antibody specifically detects R-Spondin 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| R-Spondin 1 | |
| Monoclonal | |
| Unconjugated | |
| Q2MKA7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Oncomed patent anti-RSPO1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1 | |
| RSPO1 | |
| 100 μg | |
| Signal Transduction | |
| 284654 | |
| Human | |
| Purified |