missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R-Spondin 1 Antibody (OTI11A9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73948H
This item is not returnable.
View return policy
Description
R-Spondin 1 Monoclonal specifically detects R-Spondin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| R-Spondin 1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| RSPO1 | |
| Recombinant protein fragment corresponding to amino acids 21-263 of human R-Spondin 1 (NP_001033722) produced in E.coli. | |
| 0.1 mL | |
| 284654.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI11A9 | |
| Western Blot, Immunohistochemistry | |
| CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction