Learn More
Tocris Bioscience™ (R)-ZINC 3573
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
(R)-ZINC 3573 is a Mas-related G protein-coupled receptor X2 (MRGPRX2) agonist (EC50 = 0.74 μM), which displays selectivity for MRGPRX2 over 350 other GPCRs, including MRGPRX1. (R)-ZINC 3573 induces intracellular calcium release and degranulation in LAD2 mast cells in vitro.
Negative control (S)-ZINC 3573 (Cat. No. 6352) also available.
Specifications
Specifications
| Quantity | 5 mg |
| Molecular Formula | C18H21N5 |
| Formula Weight | 307.4 |
| Product Type | Agonist |
| Purity | 98% |