Learn More
R3HDM1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41362AF750
This item is not returnable.
View return policy
Description
R3HDM1 Polyclonal antibody specifically detects R3HDM1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Western Blot | |
| Alexa Fluor 750 | |
| Primary |
| Polyclonal | |
| The epitope is recognized to a region between residue 500 to 550 of human R3H domain-containing protein 1 using the numbering given in entry Q15032 (GeneID 23518). |