Learn More
R3HDM1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41362AF750
Product Code. 18776491
  • € 669.00 / 0.10mL
Description
R3HDM1 Polyclonal antibody specifically detects R3HDM1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot
Alexa Fluor 750
Primary
Polyclonal
The epitope is recognized to a region between residue 500 to 550 of human R3H domain-containing protein 1 using the numbering given in entry Q15032 (GeneID 23518).