missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R3HDM1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41362CL1
This item is not returnable.
View return policy
Description
R3HDM1 Polyclonal antibody specifically detects R3HDM1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| R3HDM1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23518 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| R3H domain containing 1, R3H domain-containing protein 1 | |
| The epitope is recognized to a region between residue 500 to 550 of human R3H domain-containing protein 1 using the numbering given in entry Q15032 (GeneID 23518). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction