missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R9AP Antibody (D7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22470
This item is not returnable.
View return policy
Description
R9AP Monoclonal specifically detects R9AP in Mouse, Bovine samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.
Specifications
| R9AP | |
| Monoclonal | |
| 1.93 mg/mL | |
| Western Blot 1 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunocytochemistry 10 ug/ml | |
| RGS9BP | |
| Full length, bacterially expressed RGS9-1. | |
| Protein G purified | |
| RUO | |
| 388531 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| D7 | |
| Unconjugated | |
| PERRS, R9AP, regulator of G protein signaling 9 binding protein, RGS9 | |
| Mouse | |
| 57 kDa | |
| 100 μg | |
| Primary | |
| Mouse, Bovine | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido