Learn More
Epredia™ RA Lamb Slide Tray Box
Product Code. 12698476 Shop All Epredia Products
Click to view available options
:
Trays
Product Code. 12698476
Brand: Epredia™ E32
Only null left
Add to basket
Add to basket
Store and protect up to 12 microscope slide trays in this slide tray box.
  • Leatherette covering protects stored slides if box is bumped or jostled
  • Box opens and shuts using metal hinges and clasp
  • Internal wooden slide rack holds up to 48 76 x 26mm microscope slides
Please Note: Availability may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Description Capacity: 12 trays
Height (Metric) 68 mm
Length (English) Slide 3 in.
Length (Metric) 265 mm
Length (Metric) Slide 76 mm
Trays 12
Width (English) Slide 1 in.
Width (Metric) 112 mm
Width (Metric) Slide 26 mm