missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11B Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18858JF635
This item is not returnable.
View return policy
Description
RAB11B Polyclonal antibody specifically detects RAB11B in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| RAB11B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 635 | |
| GTP-binding protein YPT3, H-YPT3, MGC133246, RAB11B, member of RAS oncogene family, RAB11B, member RAS oncogene family, ras-related protein Rab-11B, YPT3 | |
| Produced in rabbits immunized with purified, recombinant Human RAB11B (Accession#: ABQ59034.1; Gly2-Cys215) | |
| 0.1 mL | |
| Cancer, GPCR, Signal Transduction | |
| 9230 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction