missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 244.00 - € 351.00
Specifications
| Antigen | RAB11B |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1- 5 uL/mg lysate |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636024
|
Novus Biologicals
NBP3-18858-100ul |
100 μg |
€ 351.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669354
|
Novus Biologicals
NBP3-18858-50ul |
50 μg |
€ 244.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB11B Polyclonal antibody specifically detects RAB11B in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| RAB11B | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, GPCR, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 9230 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1- 5 uL/mg lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| GTP-binding protein YPT3, H-YPT3, MGC133246, RAB11B, member of RAS oncogene family, RAB11B, member RAS oncogene family, ras-related protein Rab-11B, YPT3 | |
| Produced in rabbits immunized with purified, recombinant Human RAB11B (Accession#: ABQ59034.1; Gly2-Cys215) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title