missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB11FIP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142172
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise isoform 1 (NP_079427.3) and isoform 3 (NP_001002814.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Rab11-FIP1 (Rab 11 family-interacting protein 1), also known as Rab-coupling protein (RCP), is a 1283 amino acid Rab 11 effector protein. Rab11-FIP1, by interacting with Rab GTPases, is involved in the endosomal recycling process and may play a role in controlling membrane trafficking along the phagocytic pathway and during phagocytosis. Localized to the recycling endosome, the cytoplasmic membrane and phagosome membranes, Rab11-FIP1 is expressed as five isoforms produced by alternative splicing. As the most highly expressed isoform, isoform two of Rab11-FIP1 is expressed in brain, lung, testis, small intestine, spleen and heart. Isoform two of Rab11-FIP1 also has been found to form a homooligomer and is believed to interact with many Rab GTPases, including Rab 4A, Rab 11A, Rab 11B and Rab 25.
Specifications
| RAB11FIP1 | |
| Polyclonal | |
| Unconjugated | |
| RAB11FIP1 | |
| 2010200K21Rik; 4833414G05Rik; CBBM; CBP; hCG_41347; NOEL1A; Rab coupling protein; Rab effector protein; RAB11 coupling protein; RAB11 family interacting protein 1; RAB11 family interacting protein 1 (class I); rab11 family-interacting protein 1; RAB11FIP1; Rab11-FIP1; rab11fip1 {ECO:0000250; rab-coupling protein; Rab-interacting recycling protein; Rcp; RGD1562965; UniProtKB:Q6WKZ4} | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 80223 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6WKZ4 | |
| RAB11FIP1 | |
| Peptide with sequence C-EATKEAKESKKPE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction