Learn More
RAB11FIP1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16778-1-AP-150UL
 View more versions of this product
Product Code. 16846514
  • € 410.00 / 150µL
Description
Rab11-FIP1 (Rab 11 family-interacting protein 1), also known as Rab-coupling protein (RCP), is a 1283 amino acid Rab 11 effector protein. Rab11-FIP1, by interacting with Rab GTPases, is involved in the endosomal recycling process and may play a role in controlling membrane trafficking along the phagocytic pathway and during phagocytosis. Localized to the recycling endosome, the cytoplasmic membrane and phagosome membranes, Rab11-FIP1 is expressed as five isoforms produced by alternative splicing. As the most highly expressed isoform, isoform two of Rab11-FIP1 is expressed in brain, lung, testis, small intestine, spleen and heart. Isoform two of Rab11-FIP1 also has been found to form a homooligomer and is believed to interact with many Rab GTPases, including Rab 4A, Rab 11A, Rab 11B and Rab 25.
Specifications
Specifications
RAB11FIP1
Polyclonal
Unconjugated
RAB11FIP1
NOEL1A, Rab coupling protein, rab11 FIP1, RAB11FIP1, RCP
Rabbit
Antigen Affinity Chromatography
RUO
498650, 75767, 80223
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3B7T9, Q6WKZ4, Q9D620
Rab11fip1
RAB11FIP1 Fusion Protein Ag10179
150 μL
Primary
Human, Mouse, Non-human Primate, Rat