missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11FIP4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | RAB11FIP4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RAB11FIP4 Polyclonal antibody specifically detects RAB11FIP4 in Human samples. It is validated for Western BlotSpecifications
| RAB11FIP4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 84440 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ARFO2, Arfophilin-2, FIP4-Rab11, FLJ00131, KIAA1821arfophilin-2, MGC11316, MGC126566, RAB11 family interacting protein 4 (class II), rab11 family-interacting protein 4, Rab11-FIP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 338-637 of human RAB11FIP4 (NP_116321.2). NDITEKVSFLEKKVTELENDSLTNGDLKSKLKQENTQLVHRVHELEEMVKDQETTAEQALEEEARRHREAYGKLEREKATEVELLNARVQQLEEENTELRTTVTRLKSQTEKLDEERQRMSDRLEDTSLRLKDEMDLYKRMMDKLRQNRLEFQKEREATQELIEDLRKELEHLQMYKLDCERPGRGRSASSGLGEFNARAREVELEHEVKRLKQENYKLRDQNDDLNGQILSLSLYEAKNLFAAQTKAQSLAAEIDTASRDELMEALKEQEEINFRLRQYMDKIILAILDHNPSILEIKH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title