missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB11FIP4 Monoclonal Antibody (OTI13B5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526978
This item is not returnable.
View return policy
Description
RAB11FIP4 Monoclonal Antibody for Western Blot, IHC (P)
Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP4 is one of many proteins that interact with and regulate Rab GTPases.
Specifications
| RAB11FIP4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86YS3 | |
| RAB11FIP4 | |
| Human recombinant protein fragment corresponding to amino acids 91-177 of RAB11FIP4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI13B5 | |
| Unconjugated | |
| RAB11FIP4 | |
| A730072L08Rik; ARFO2; Arfophilin-2; FIP4-Rab11; Kiaa1821; mKIAA1821; mRab11-FIP4A; RAB11 family interacting protein 4; RAB11 family interacting protein 4 (class II); rab11 family-interacting protein 4; Rab11fip4; RAB11-FIP4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84440 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction